Assay Details
| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968266.1 | XP_017823755.1 | 2 - 3 | 1410 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHST7| Gene Symbol: CHST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968266.1 | XP_017823755.1 | 2 - 3 | 1410 | 106 | |
Set Membership: |
|