Assay Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000459.2 | XP_008998707.1 | 4 - 5 | 1090 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGMAT| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000459.2 | XP_008998707.1 | 4 - 5 | 1090 | 78 | |
Set Membership: |
|