Assay Details
| Gene Symbol: CDKN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753918.4 | XP_002753964.1 | 5 - 6 | 547 | 80 | |
| XM_009006052.2 | XP_009004300.1 | 4 - 5 | 449 | 80 | ||
More Information
Set Membership: |
|

CDKN3
Genomic Map