Assay Details
| Gene Symbol: KBTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998183.2 | XP_008996431.1 | 2 | 3696 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KBTBD6| Gene Symbol: KBTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998183.2 | XP_008996431.1 | 2 | 3696 | 138 | |
Set Membership: |
|