Assay Details
| Gene Symbol: LOC100407631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991622.2 | XP_008989870.1 | 3 - 4 | 775 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008991622.2 transcripts.
Set Membership: |
|

LOC100407631
Genomic Map