Assay Details
| Gene Symbol: ZNRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746299.4 | XP_002746345.2 | 4 - 5 | 505 | 91 | |
| XM_008994089.2 | XP_008992337.1 | 4 - 5 | 475 | 91 | ||
More Information
Set Membership: |
|

ZNRD1
Genomic Map