Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009007952.2 | XP_009006200.1 | 4 | 2692 | 91 | |
XM_009007954.2 | XP_009006202.1 | 2 | 2560 | 91 | ||
XM_009007955.2 | XP_009006203.1 | 3 | 2612 | 91 | ||
XM_017978024.1 | XP_017833513.1 | 5 | 2758 | 91 | ||
XM_017978025.1 | XP_017833514.1 | 5 | 2753 | 91 | ||
XM_017978026.1 | XP_017833515.1 | 6 | 1883 | 91 | ||
XM_017978027.1 | XP_017833516.1 | 3 | 2640 | 91 | ||
XM_017978028.1 | XP_017833517.1 | 4 | 2678 | 91 | ||
XM_017978030.1 | XP_017833519.1 | 4 | 2673 | 91 | ||
GenBank mRNA | EF215150.1 | - | 1 | 121 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
