Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748531.4 | XP_002748577.3 | 2 - 3 | 323 | 66 | |
| XM_008997505.1 | XP_008995753.1 | 2 - 3 | 442 | 66 | ||
| XM_008997506.2 | XP_008995754.1 | 2 - 3 | 314 | 66 | ||
| XM_008997507.2 | XP_008995755.1 | 2 - 3 | 694 | 66 | ||
More Information
Set Membership: |
|

ORMDL3
Genomic Map