Assay Details
| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006215.2 | XP_009004463.1 | 4 - 5 | 560 | 78 | |
| XM_009006216.2 | XP_009004464.1 | 4 - 5 | 655 | 78 | ||
More Information
Set Membership: |
|

RDH11
Genomic Map