Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749806.3 | XP_002749852.1 | 7 - 8 | 855 | 64 | |
| XM_017973742.1 | XP_017829231.1 | 8 - 9 | 689 | 64 | ||
More Information
Set Membership: |
|

ZFAND2B
Genomic Map