Assay Details
| Gene Symbol: HUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751405.3 | XP_002751451.1 | 3 - 4 | 505 | 58 | |
| XM_017975170.1 | XP_017830659.1 | 3 - 4 | 505 | 58 | ||
| XM_017975171.1 | XP_017830660.1 | 4 - 5 | 535 | 58 | ||
More Information
Set Membership: |
|

HUS1
Genomic Map