Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760794.3 | XP_002760840.1 | 4 - 5 | 1043 | 82 | |
| XM_008985642.2 | XP_008983890.1 | 5 - 6 | 1166 | 82 | ||
| XM_017966652.1 | XP_017822141.1 | 4 - 5 | 1527 | 82 | ||
| XM_017966653.1 | XP_017822142.1 | 3 - 4 | 1406 | 82 | ||
| XM_017966654.1 | XP_017822143.1 | 5 - 6 | 1221 | 82 | ||
| XM_017966655.1 | XP_017822144.1 | 4 - 5 | 1524 | 82 | ||
| XM_017966656.1 | XP_017822145.1 | 4 - 5 | 1529 | 82 | ||
More Information
Set Membership: |
|

ZNF496
Genomic Map