Assay Details
| Gene Symbol: URGCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002102.2 | XP_009000350.1 | 2 - 3 | 516 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009002102.2 transcripts.
Set Membership: |
|

URGCP
Genomic Map