Assay Details
| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972234.1 | XP_017827723.1 | 7 - 8 | 1010 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHMT1| Gene Symbol: SHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972234.1 | XP_017827723.1 | 7 - 8 | 1010 | 61 | |
Set Membership: |
|