Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761208.4 | XP_002761254.1 | 2 - 3 | 1961 | 69 | |
| XM_008986274.2 | XP_008984522.1 | 2 - 3 | 193 | 69 | ||
| XM_017967027.1 | XP_017822516.1 | 2 - 3 | 188 | 69 | ||
More Information
Set Membership: |
|

SDR42E1
Genomic Map