Assay Details
| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733159.2 | XP_003733207.1 | 2 - 3 | 753 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FKBP7| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733159.2 | XP_003733207.1 | 2 - 3 | 753 | 65 | |
Set Membership: |
|