Assay Details
| Gene Symbol: SPOCK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992478.2 | XP_008990726.1 | 8 - 9 | 680 | 65 | |
| XM_008992479.2 | XP_008990727.1 | 8 - 9 | 687 | 65 | ||
| XM_008992480.1 | XP_008990728.1 | 7 - 8 | 708 | 65 | ||
| XM_008992481.2 | XP_008990729.1 | 7 - 8 | 671 | 65 | ||
| XM_008992482.2 | XP_008990730.1 | 6 - 7 | 662 | 65 | ||
| XM_017970160.1 | XP_017825649.1 | 7 - 8 | 573 | 65 | ||
More Information
Set Membership: |
|

SPOCK3
Genomic Map