Assay Details
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982234.2 | XP_008980482.1 | 3 - 4 | 1440 | 62 | |
XM_008982235.2 | XP_008980483.1 | 2 - 3 | 603 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
