Assay Details
| Gene Symbol: UQCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747246.2 | XP_002747292.1 | 8 - 9 | 827 | 100 | |
| XM_002747247.3 | XP_002747293.1 | 7 - 8 | 601 | 100 | ||
More Information
Set Membership: |
|

UQCC
Genomic Map