Assay Details
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963341.1 | XP_017818830.1 | 5 - 6 | 1193 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963341.1 | XP_017818830.1 | 5 - 6 | 1193 | 93 |
Set Membership: |
![]() ![]() ![]() |