Assay Details
| Gene Symbol: LOC108593534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977232.1 | XP_017832721.1 | 3 - 4 | 838 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC108593534| Gene Symbol: LOC108593534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977232.1 | XP_017832721.1 | 3 - 4 | 838 | 97 | |
Set Membership: |
|