Assay Details
| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734959.3 | XP_003735007.2 | 2 - 3 | 600 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBSPON| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734959.3 | XP_003735007.2 | 2 - 3 | 600 | 92 | |