Assay Details
| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734959.3 | XP_003735007.2 | 4 - 5 | 869 | 89 | |
| XM_008982770.2 | XP_008981018.1 | 5 - 6 | 838 | 89 | ||
More Information
Set Membership: |
|

SBSPON
Genomic Map