Assay Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989878.2 | XP_008988126.1 | 17 | 3150 | 101 | |
| XM_008989880.2 | XP_008988128.2 | 16 | 3233 | 101 | ||
| XM_008989881.2 | XP_008988129.1 | 16 | 3086 | 101 | ||
| XM_017968666.1 | XP_017824155.1 | 17 | 2945 | 101 | ||
| XM_017968667.1 | XP_017824156.1 | 16 | 3832 | 101 | ||
More Information
Set Membership: |
|

ZNF280C
Genomic Map