Assay Details
| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754245.4 | XP_002754291.1 | 5 | 1968 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINA10| Gene Symbol: SERPINA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754245.4 | XP_002754291.1 | 5 | 1968 | 99 | |
Set Membership: |
|