Assay Details
| Gene Symbol: CEACAM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967660.1 | XP_017823149.1 | 3 - 4 | 760 | 88 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEACAM19| Gene Symbol: CEACAM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967660.1 | XP_017823149.1 | 3 - 4 | 760 | 88 | |