Assay Details
| Gene Symbol: PLAU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756216.3 | XP_002756262.1 | 10 - 11 | 1266 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLAU| Gene Symbol: PLAU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756216.3 | XP_002756262.1 | 10 - 11 | 1266 | 67 | |
Set Membership: |
|