Assay Details
| Gene Symbol: LOC100403511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759822.4 | XP_002759868.1 | 12 - 13 | 1269 | 91 | |
| XM_008984197.2 | XP_008982445.1 | 12 - 13 | 1275 | 91 | ||
| XM_008984200.1 | XP_008982448.1 | 11 - 12 | 1167 | 91 | ||
| XM_017966197.1 | XP_017821686.1 | 10 - 11 | 1029 | 91 | ||
More Information
Set Membership: |
|

LOC100403511
Genomic Map