Assay Details
| Gene Symbol: LOC100403511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759822.4 | XP_002759868.1 | 9 - 10 | 984 | 96 | |
| XM_008984197.2 | XP_008982445.1 | 9 - 10 | 990 | 96 | ||
| XM_008984200.1 | XP_008982448.1 | 8 - 9 | 882 | 96 | ||
| XM_017966197.1 | XP_017821686.1 | 7 - 8 | 744 | 96 | ||
More Information
Set Membership: |
|

LOC100403511
Genomic Map