Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755288.4 | XP_002755334.1 | 2 - 3 | 1328 | 79 | |
| XM_009008150.2 | XP_009006398.1 | 3 - 4 | 853 | 79 | ||
| XM_009008151.2 | XP_009006399.1 | 2 - 3 | 752 | 79 | ||
More Information
Set Membership: |
|

CLP1
Genomic Map