Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002742737.4 | XP_002742783.2 | 7 - 8 | 3533 | 63 | |
| XM_002742738.3 | XP_002742784.1 | 8 - 9 | 2462 | 63 | ||
| XM_008993373.2 | XP_008991621.1 | 8 - 9 | 2438 | 63 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map