Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987788.1 | XP_008986036.1 | 1 - 2 | 286 | 86 | |
| XM_008987789.1 | XP_008986037.1 | 1 - 2 | 286 | 86 | ||
More Information
Set Membership: |
|

THAP8
Genomic Map