Assay Details
| Gene Symbol: BTN3A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971580.1 | XP_017827069.1 | 7 - 8 | 1037 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTN3A3| Gene Symbol: BTN3A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971580.1 | XP_017827069.1 | 7 - 8 | 1037 | 90 | |
Set Membership: |
|