Assay Details
| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978868.1 | XP_017834357.1 | 2 - 3 | 325 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHKG2| Gene Symbol: PHKG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978868.1 | XP_017834357.1 | 2 - 3 | 325 | 72 | |
Set Membership: |
|