Assay Details
| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747285.3 | XP_002747331.2 | 2 - 3 | 552 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PROCR| Gene Symbol: PROCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747285.3 | XP_002747331.2 | 2 - 3 | 552 | 95 | |
Set Membership: |
|