Assay Details
| Gene Symbol: ABI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733010.3 | XP_003733058.1 | 3 - 4 | 957 | 67 | |
| XM_008997400.2 | XP_008995648.1 | 3 - 4 | 602 | 67 | ||
| XM_008997401.2 | XP_008995649.1 | 2 - 3 | 316 | 67 | ||
More Information
Set Membership: |
|

ABI3
Genomic Map