Assay Details
| Gene Symbol: RANBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993940.2 | XP_008992188.2 | 12 - 13 | 1409 | 78 | |
| XM_017970905.1 | XP_017826394.1 | 11 - 12 | 1393 | 78 | ||
More Information
Set Membership: |
|

RANBP9
Genomic Map