Assay Details
| Gene Symbol: ALS2CR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973177.1 | XP_017828666.1 | 13 - 14 | 3496 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALS2CR12| Gene Symbol: ALS2CR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973177.1 | XP_017828666.1 | 13 - 14 | 3496 | 68 | |
Set Membership: |
|