Assay Details
| Gene Symbol: OGFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753131.3 | XP_002753177.1 | 4 - 5 | 464 | 92 | |
| XM_017976470.1 | XP_017831959.1 | 3 - 4 | 350 | 92 | ||
| XM_017976471.1 | XP_017831960.1 | 4 - 5 | 464 | 92 | ||
More Information
Set Membership: |
|

OGFOD2
Genomic Map