Assay Details
| Gene Symbol: NFYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974600.1 | XP_017830089.1 | 7 - 8 | 1225 | 76 | |
| GenBank mRNA | EF215005.1 | - | 1 | 237 | 76 | |
More Information
Set Membership: |
|

NFYC
Genomic Map