Assay Details
| Gene Symbol: DCLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806645.4 | XP_002806691.2 | 11 - 12 | 2367 | 70 | |
| XM_008992612.2 | XP_008990860.1 | 11 - 12 | 1612 | 70 | ||
| XM_008992613.2 | XP_008990861.1 | 10 - 11 | 2321 | 70 | ||
| XM_008992614.2 | XP_008990862.1 | 10 - 11 | 1561 | 70 | ||
| XM_017970210.1 | XP_017825699.1 | 11 - 12 | 1615 | 70 | ||
More Information
Set Membership: |
|

DCLK2
Genomic Map