Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734679.3 | XP_003734727.1 | 5 - 6 | 600 | 65 | |
| XM_008980142.2 | XP_008978390.1 | 5 - 6 | 589 | 65 | ||
| XM_008980143.2 | XP_008978391.1 | 5 - 6 | 580 | 65 | ||
| XM_008980145.2 | XP_008978393.1 | 5 - 6 | 566 | 65 | ||
| XM_008980146.2 | XP_008978394.1 | 5 - 6 | 578 | 65 | ||
More Information
Set Membership: |
|

CHST10
Genomic Map