Assay Details
| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757156.3 | XP_002757202.1 | 1 - 2 | 271 | 61 | |
| XM_008979663.2 | XP_008977911.1 | 1 - 2 | 273 | 61 | ||
More Information
Set Membership: |
|

GAREM1
Genomic Map