Assay Details
| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757156.3 | XP_002757202.1 | 5 - 6 | 1880 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAREM1| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757156.3 | XP_002757202.1 | 5 - 6 | 1880 | 76 | |
Set Membership: |
|