Assay Details
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752030.4 | XP_002752076.2 | 1 - 2 | 301 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
