Assay Details
| Gene Symbol: PSMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751176.3 | XP_002751222.1 | 6 - 7 | 574 | 75 | |
| XM_009001839.2 | XP_009000087.1 | 5 - 6 | 491 | 75 | ||
More Information
Set Membership: |
|

PSMA5
Genomic Map