Assay Details
| Gene Symbol: C18H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760008.3 | XP_002760054.1 | 3 - 4 | 503 | 96 | |
| XM_008984475.2 | XP_008982723.1 | 3 - 4 | 565 | 96 | ||
More Information
Set Membership: |
|

C18H1orf189
Genomic Map