Assay Details
| Gene Symbol: FBXO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009737.2 | XP_009007985.1 | 3 - 4 | 1131 | 66 | |
| XM_009009743.2 | XP_009007991.1 | 3 - 4 | 523 | 66 | ||
More Information
Set Membership: |
|

FBXO7
Genomic Map