Assay Details
| Gene Symbol: CSGALNACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756099.3 | XP_002756145.1 | 5 - 6 | 1534 | 89 | |
| XM_009009488.1 | XP_009007736.1 | 4 - 5 | 1317 | 89 | ||
| XM_017978883.1 | XP_017834372.1 | 5 - 6 | 1534 | 89 | ||
| XM_017978884.1 | XP_017834373.1 | 2 - 3 | 314 | 89 | ||
More Information
Set Membership: |
|

CSGALNACT2
Genomic Map