Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017964056.1 | XP_017819545.1 | 6 - 7 | 897 | 80 | |
| XM_017964057.1 | XP_017819546.1 | 6 - 7 | 793 | 80 | ||
More Information
Set Membership: |
|

KCNIP3
Genomic Map